1H,1H,9H-Hexadecafluor-1-nonanol ≥97%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluor-1-nonanol, 1H,1H,9H-Perfluor-1-nonanol
B21407.22
B21407.22EA
266
EUR
B21407.22
1H,1H,9H-Hexadecafluor-1-nonanol ≥97%
1H,1H,9H-Hexadecafluor-1-nonanol
Purity: 97%
CAS No.: 376-18-1
Molecular Formula: H(CF2)8CH2OH
Formula Weight: 432.11
Melting Point: 55-60°
Boiling Point: 157-158°/200mm
MDL No.: MFCD00039629
Formel:
C₉H₄F₁₆O Molecular Weight: 432,1 g/mol Siedepunkt: 157…158 °C (200 mmHg) Schmelzpunkt: 55…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039629 CAS-Nummer: 376-18-1 EINECS: 206-806-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...