2-Noninsäuremethylester ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl-2-noninoat

B21409.14
B21409.14EA 50.2 EUR
B21409.14
2-Noninsäuremethylester ≥98%
2-Nonynoic acid methyl ester

Purity: 98% CAS No.: 111-80-8 Molecular Formula: C10H16O2 Formula Weight: 168.24 Flash Point: 104°(219°F) Refractive Index: 1.4485 Odour: Floral Boiling Point: 121°/20mm Beilstein Reference: 1759870 MDL No.: MFCD00009547

Formel: C₁₀H₁₆O₂
Molecular Weight: 168,24 g/mol
Siedepunkt: 121 °C (20 mmHg)
Dichte: 0,915
Flammpunkt: 104 °C (219 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009547
CAS-Nummer: 111-80-8
EINECS: 203-909-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR