cis-1,2,3,6-Tetrahydrophthalsäureanhydrid 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
cis-4-Cyclohexene-1,2-dicarboxylic anhydride
B21431.30
B21431.30EA
26.6
EUR
B21431.30
cis-1,2,3,6-Tetrahydrophthalsäureanhydrid 95%
cis-1,2,3,6-Tetrahydrophthalsäureanhydrid
Purity: 95%
CAS No.: 935-79-5
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 97-103°
Beilstein Reference: 82341
Formel:
C₈H₈O₃ Molecular Weight: 152,15 g/mol Schmelzpunkt: 97…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005916 CAS-Nummer: 935-79-5 EINECS: 213-308-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...