4-Brom-3,5-dimethoxybenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B21449.06 B21449.14
B21449.06EA 27.6 EUR
B21449.06 B21449.14
4-Brom-3,5-dimethoxybenzoesäure ≥98%
4-Bromo-3,5-dimethoxybenzoic acid

Purity: 98% CAS No.: 56518-42-4 Molecular Formula: C9H9BrO4 Formula Weight: 261.07 Melting Point: 254-258° MDL No.: MFCD01632140

Formel: C₉H₉BrO₄
Schmelzpunkt: 254…258 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632140
CAS-Nummer: 56518-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR