3-Methylanisol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Kresolmethylether
B21455.18
B21455.30
B21455.18EA
35.9
EUR
B21455.18
B21455.30
3-Methylanisol ≥99%
3-Methylanisol
Purity: 99%
CAS No.: 100-84-5
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 54°(129°F)
Refractive Index: 1.5130
Odour: Pleasant
Boiling Point: 175-176°
Beilstein Reference: 907023
UN No.: 3271
MDL No.: MFCD00008395
Formel:
C₈H₁₀O Molecular Weight: 122.17 g/mol Siedepunkt: 175…176 °C Schmelzpunkt: –47 °C Dichte: 0.969 Flammpunkt: 54 °C (129 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008395 CAS-Nummer: 100-84-5 EINECS: 202-893-4 UN: 3271 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...