5-Fluoranthranilsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B21458.03 B21458.06 B21458.14
B21458.03EA 61.5 EUR
B21458.03 B21458.06 B21458.14
5-Fluoranthranilsäure ≥98%
5-Fluoranthranilsäure

Purity: 98+% CAS No.: 446-08-2 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Melting Point: 182-184° Beilstein Reference: 2803664

Formel: C₇H₆FNO₂
Siedepunkt: 305 °C (1013 hPa)
Schmelzpunkt: 182…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00055566
CAS-Nummer: 446-08-2
EINECS: 207-159-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR