Propylbenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 103-65-1
Molecular Formula: C9H12
Formula Weight: 120.20
Flash Point: 47°(116°F)
Refractive Index: 1.4910
Melting Point: -100°
Boiling Point: 159-160°
Beilstein Reference: 1903006
Merck Reference: 14,7844
UN No.: 1993
MDL No.: MFCD00009377
Formel:
C₉H₁₂ Siedepunkt: 159…160 °C Schmelzpunkt: –100 °C Dichte: 0,862 Flammpunkt: 47 °C (116 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009377 CAS-Nummer: 103-65-1 EINECS: 203-132-9 UN: 1993 ADR: 3,III Merck Index: 14,7844 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...