4-Butoxybenzoesäure ≥98%
Lieferant: Thermo Fisher Scientific
B21506.14
B21506.22
B21506.14EA
68.7
EUR
B21506.14
B21506.22
4-Butoxybenzoesäure ≥98%
4-Butoxybenzoesäure
Purity: 98%
CAS No.: 1498-96-0
Molecular Formula: C11H14O3
Formula Weight: 194.23
Melting Point: 146-147° (C-N)
MDL No.: MFCD00002546
Formel:
C₁₁H₁₄O₃ Siedepunkt: 319 °C (1013 hPa) Schmelzpunkt: 146…147 °C (C-nematisch) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002546 CAS-Nummer: 1498-96-0 EINECS: 216-104-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...