Xylene cyanole FF ≥70% (Farbstoffgehalt)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Acid Blue 147, Hydrogen-4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-yliden]methyl]benzol-1,3-disulfonatnatrium
B21530.09
B21530.18
B21530.09EA
48.9
EUR
B21530.09
B21530.18
Xylene cyanole FF ≥70% (Farbstoffgehalt)
Xylencyanol FF
CAS No.: 2650-17-1
Molecular Formula: C25H27N2NaO6S2
Formula Weight: 538.61
Melting Point: ca 295° dec.
MDL No.: MFCD00019481
Formel:
C₂₅H₂₇N₂NaO₆S₂ Molecular Weight: 538.62 g/mol Schmelzpunkt: ∼ 295 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040657 CAS-Nummer: 2650-17-1 EINECS: 220-167-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...