2,2,4,4-Tetrafluoro-1,3-dithietane 97%

Lieferant: Thermo Fisher Scientific
Achtung

B21620.09
B21620.09EA 29.1 EUR
B21620.09
2,2,4,4-Tetrafluoro-1,3-dithietane 97%
2,2,4,4-Tetrafluoro-1,3-dithietane

Purity: 97% CAS No.: 1717-50-6 Molecular Formula: C2F4S2 Formula Weight: 164.14 Flash Point: 36°(96°F) Refractive Index: 1.393 Odour: Unpleasant Boiling Point: 47-49° UN No.: UN1993 MDL No.: MFCD00053538

Formel: C₂F₄S₂
Molecular Weight: 164.14 g/mol
Siedepunkt: 47…49 °C
Dichte: 1.60
Flammpunkt: 36 °C (96 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00053538
CAS-Nummer: 1717-50-6
EINECS: 217-001-9
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR