3-(2-Furyl)acrylsäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

B21653.14 B21653.22 B21653.30
B21653.14EA 40.2 EUR
B21653.14 B21653.22 B21653.30
3-(2-Furyl)acrylsäure ≥99%
3-(2-Furyl)acrylsäure

Purity: 99% CAS No.: 539-47-9 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 140-144° Boiling Point: 286° Merck Reference: 14,4297 MDL No.: MFCD00003257

Formel: C₇H₆O₃
Siedepunkt: 286 °C
Schmelzpunkt: 140…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003257
CAS-Nummer: 539-47-9
EINECS: 208-718-8
Merck Index: 14,4297

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR