3-(Trifluormethyl)phenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(Trifluormethyl)benzolboronsäure, Dihydroxy(3-(trifluormethyl)phenyl)boran, (3-(Trifluormethyl)phenyl)metaborsäure

B21661.03 B21661.06 B21661.14
B21661.03EA 32.8 EUR
B21661.03 B21661.06 B21661.14
3-(Trifluormethyl)phenylboronsäure ≥98%
3-(Trifluormethyl)phenylboronsäure

Purity: 98% CAS No.: 1423-26-3 Molecular Formula: C7H6BF3O2 Formula Weight: 189.93 Melting Point: 162-166° Beilstein Reference: 6084746

Formel: C₇H₆BF₃O₂
Molecular Weight: 189.93 g/mol
Schmelzpunkt: 162…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151854
CAS-Nummer: 1423-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR