Tris(2-hydroxyethyl)-1,3,5-triazintrion ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B21697.0B B21697.18 B21697.30
B21697.0BEA 443 EUR
B21697.0B B21697.18 B21697.30
Tris(2-hydroxyethyl)-1,3,5-triazintrion ≥97%
Tris(2-hydroxyethyl)-1,3,5-triazintrion

Purity: 97% CAS No.: 839-90-7 Molecular Formula: C9H15N3O6 Formula Weight: 261.24 Melting Point: 136-140° MDL No.: MFCD00003549

Formel: C₉H₁₅N₃O₆
Siedepunkt: 527 °C (1013 hPa)
Schmelzpunkt: 136…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003549
CAS-Nummer: 839-90-7
EINECS: 212-660-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR