9-Fluorenone hydrazone 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(9H-Fluoren-9-ylidene)hydrazine
B21706.14
B21706.14EA
80.1
EUR
B21706.14
9-Fluorenone hydrazone 97%
9-Fluorenone hydrazone
Purity: 98+%
CAS No.: 13629-22-6
Molecular Formula: C13H10N2
Formula Weight: 194.24
Melting Point: 150-154°
Beilstein Reference: 1910575
MDL No.: MFCD00016357
Formel:
C₁₃H₁₀N₂ Molecular Weight: 194.24 g/mol Schmelzpunkt: 150…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016357 CAS-Nummer: 13629-22-6 EINECS: 237-116-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...