Phenolrot

Lieferant: Thermo Fisher Scientific
Achtung

B21710.09 B21710.18 B21710.30
B21710.09EA 33 EUR
B21710.09 B21710.18 B21710.30
Phenolrot
Phenolrot

CAS No.: 143-74-8 Molecular Formula: C19H14O5S Formula Weight: 354.38 Odour: Odourless Boiling Point: 300° Beilstein Reference: 326470 Merck Reference: 14,7247 MDL No.: MFCD00003552

Formel: C₁₉H₁₄O₅S
Siedepunkt: >300 °C
Schmelzpunkt: > 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003552
CAS-Nummer: 143-74-8
EINECS: 205-609-7
Merck Index: 14,7247

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR