trans-4-(Aminomethyl)cyclohexancarbonsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: HAKU, AMCA, TAMCHA, trans-4-(aminomethyl)cyclohexane-1-carboxylic acid, Tranexamic acid, AMCHA

B21742.09 B21742.18 B21742.30
B21742.09EA 52.3 EUR
B21742.09 B21742.18 B21742.30
trans-4-(Aminomethyl)cyclohexancarbonsäure 97%
trans-4-(Aminomethyl)cyclohexanecarboxylic acid

Purity: 97% CAS No.: 1197-18-8 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Melting Point: 300° Beilstein Reference: 2207452 Merck Reference: 14,9569 MDL No.: MFCD00001466

Formel: C₈H₁₅NO₂
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001466
CAS-Nummer: 1197-18-8
EINECS: 214-818-2
Merck Index: 14,9569

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR