4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%

Lieferant: Thermo Fisher Scientific

Synonyme: BOC-L-Aspartic acid 4-benzylester, 4-Benzyl N-Boc-L-aspartate, Boc-Asp(Obzl)-OH, Boc-Asp(OBzl)-OH, N-Boc-L-aspartic Acid 4-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Benzyl Ester

B21758.06
B21758.06EA 28.6 EUR
B21758.06
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate

Purity: 98% CAS No.: 7536-58-5 Molecular Formula: C16H21NO6 Formula Weight: 323.35 Melting Point: 110-112° MDL No.: MFCD00065564

Formel: C₁₆H₂₁NO₆
Molecular Weight: 323.35 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065564
CAS-Nummer: 7536-58-5
EINECS: 231-406-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR