7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt 99%
Lieferant: Thermo Scientific
Synonyme:
Ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate
B21767.MF
B21767.MFEA
633
EUR
B21767.MF
7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt 99%
7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt
Purity: 99%
CAS No.: 84806-27-9
Molecular Formula: C6H6FN3O4S
Formula Weight: 235.19
Storage Temperature: 2°C to 8°C
Melting Point: ca. 285-295°C dec.
MDL No.: MFCD02682109
Formel:
C₆H₆FN₃O₄S Molecular Weight: 235,189 g/mol Schmelzpunkt: ∼285…295 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD02682109 CAS-Nummer: 84806-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...