3-n-Hexylpyridine ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B21775.88
B21775.88EA 40.6 EUR
B21775.88
3-n-Hexylpyridine ≥98%
3-n-Hexylpyridine

Purity: 98+% CAS No.: 6311-92-8 Molecular Formula: C11H17N Formula Weight: 163.27 MDL No.: MFCD02683071

Formel: C₁₁H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD02683071
CAS-Nummer: 6311-92-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR