2-Pentyl-1,3-propandiol ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Amyl-1,3-propandiol

B21800.06
B21800.06EA 91 EUR
B21800.06
2-Pentyl-1,3-propandiol ≥97%
2-Pentyl-1,3-propanediol

Purity: 97% CAS No.: 25462-23-1 Molecular Formula: CH3(CH2)4CH(CH2OH)2 Formula Weight: 146.23 MDL No.: MFCD00801123

Formel: C₈H₁₈O₂
Molecular Weight: 146.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00801123
CAS-Nummer: 25462-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR