2,6-Dimethoxy-p-kresol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,5-Dimethoxy-4-hydroxytoluol, 2,6-Dimethoxy-4-methylphenol, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzol

B21801.14 B21801.22
B21801.14EA 63.6 EUR
B21801.14 B21801.22
2,6-Dimethoxy-p-kresol ≥97%
2,6-Dimethoxy-p-kresol

Purity: 97% CAS No.: 6638-05-7 Molecular Formula: C9H12O3 Formula Weight: 168.19 Flash Point: 110°(230°F) Odour: Phenol-like Boiling Point: 144-146°/12mm MDL No.: MFCD00017289

Formel: C₉H₁₂O₃
Molecular Weight: 168.19 g/mol
Siedepunkt: 144…146 °C (12 mmHg)
Schmelzpunkt: 36…42 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00017289
CAS-Nummer: 6638-05-7
EINECS: 229-641-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR