2,6-Dimethoxy-p-kresol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,5-Dimethoxy-4-hydroxytoluol, 2,6-Dimethoxy-4-methylphenol, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzol
B21801.14
B21801.22
B21801.14EA
63.6
EUR
B21801.14
B21801.22
2,6-Dimethoxy-p-kresol ≥97%
2,6-Dimethoxy-p-kresol
Purity: 97%
CAS No.: 6638-05-7
Molecular Formula: C9H12O3
Formula Weight: 168.19
Flash Point:
Formel:
C₉H₁₂O₃ Molecular Weight: 168.19 g/mol Siedepunkt: 144…146 °C (12 mmHg) Schmelzpunkt: 36…42 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017289 CAS-Nummer: 6638-05-7 EINECS: 229-641-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...