Sodium ethanesulphonate, (max. 2% H₂O) 98%

Lieferant: Thermo Fisher Scientific
Achtung

B21824.06 B21824.14
B21824.06EA 67.1 EUR
B21824.06 B21824.14
Sodium ethanesulphonate, (max. 2% H₂O) 98%
Sodium ethanesulphonate

Purity: 98% CAS No.: 5324-47-0 Molecular Formula: CH3CH2SO3Na Formula Weight: 132.11 Melting Point: ca 270° MDL No.: MFCD00066498

Formel: C₂H₆O₃S
Schmelzpunkt: ∼ 270 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066498
CAS-Nummer: 5324-47-0
EINECS: 226-194-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR