3-Aminobenzolsulfonamid ≥97%

Lieferant: Thermo Scientific

Synonyme: Anilin-3-sulfonsäureamid, 3-Aminobenzolsulfonsäureamid

B21857.06
B21857.06EA 25.6 EUR
B21857.06
3-Aminobenzolsulfonamid ≥97%
3-Aminobenzolsulfonamid

Purity: 97+% CAS No.: 98-18-0 Molecular Formula: C6H8N2O2S Formula Weight: 172.21 Melting Point: 139-142° MDL No.: MFCD00035781

Formel: C₆H₈N₂O₂S
Molecular Weight: 172,21 g/mol
Schmelzpunkt: 139…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00035781
CAS-Nummer: 98-18-0
EINECS: 202-646-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR