4-(4-Hydroxyphenyl)benzonitril ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4'-Hydroxy-4-biphenylcarbonitrile, 4'-Hydroxybiphenyl-4-carbonitrile, 4'-Hydroxy-[1,1'-biphenyl]-4-carbonitrile

B21864.06
B21864.06EA 73 EUR
B21864.06
4-(4-Hydroxyphenyl)benzonitril ≥99%
4-(4-Hydroxyphenyl)benzonitril

Purity: 99% CAS No.: 19812-93-2 Molecular Formula: C13H9NO Formula Weight: 195.22 Melting Point: 196-199° UN No.: 3439 MDL No.: MFCD00059625

Formel: C₁₃H₉NO
Molecular Weight: 195,22 g/mol
Schmelzpunkt: 196…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059625
CAS-Nummer: 19812-93-2
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR