(R)-(+)-1,2-Epoxypropan ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

B21884.06 B21884.14 B21884.22
B21884.06EA 76 EUR
B21884.06 B21884.14 B21884.22
(R)-(+)-1,2-Epoxypropan ≥99%
(R)-(+)-1,2-Epoxypropan

Purity: 99% CAS No.: 15448-47-2 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -37°(-35°F) Odour: Ether-like Boiling Point: 34-35° Beilstein Reference: 79764 Merck Reference: 14,7856 UN No.: 1280 MDL No.: MFCD00066211

Formel: C₃H₆O
Siedepunkt: 34…35 °C
Dichte: 0,829
Flammpunkt: –37 °C (–35 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066211
CAS-Nummer: 15448-47-2
UN: 1280
ADR: 3,I
Merck Index: 14,7856

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR