N-Acetylglycin (Ac-Gly-OH) ≥99%
Lieferant: Thermo Fisher Scientific
B21887.22
B21887.36
B21887.22EA
30.3
EUR
B21887.22
B21887.36
N-Acetylglycin (Ac-Gly-OH) ≥99%
N-Acetylglycin (Ac-Gly-OH)
Purity: 99%
CAS No.: 543-24-8
Molecular Formula: CH3CONHCH2CO2H
Formula Weight: 117.10
Melting Point: ca 210° dec.
Beilstein Reference: 774114
Merck Reference: 14,80
MDL No.: MFCD00004275
Formel:
C₄H₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004275 CAS-Nummer: 543-24-8 EINECS: 208-839-6 Merck Index: 14,80 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...