N-Acetylglycin (Ac-Gly-OH) ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid

B21887.22 B21887.36
B21887.22EA 30.3 EUR
B21887.22 B21887.36
N-Acetylglycin (Ac-Gly-OH) ≥99%
N-Acetylglycin (Ac-Gly-OH)

Purity: 99% CAS No.: 543-24-8 Molecular Formula: CH3CONHCH2CO2H Formula Weight: 117.10 Melting Point: ca 210° dec. Beilstein Reference: 774114 Merck Reference: 14,80 MDL No.: MFCD00004275

Formel: C₄H₇NO₃
Schmelzpunkt: ∼ 210 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004275
CAS-Nummer: 543-24-8
EINECS: 208-839-6
Merck Index: 14,80

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR