2-(Trimethylsilyl)thiazol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B21903.14 B21903.06
B21903.14EA 664 EUR
B21903.14 B21903.06
2-(Trimethylsilyl)thiazol ≥97%
2-(Trimethylsilyl)thiazole

Purity: 97% CAS No.: 79265-30-8 Molecular Formula: C6H11NSSi Formula Weight: 157.31 Storage Temperature: 2°C to 8°C Flash Point: 57°(135°F) Refractive Index: 1.4970 Boiling Point: 53-55°/10mm UN No.: 1993 MDL No.: MFCD00066274

Formel: C₆H₁₁NSSi
Siedepunkt: 53…55 °C (10 mmHg)
Dichte: 0,985
Flammpunkt: 57 °C (135 °F)
MDL: MFCD00066274
CAS-Nummer: 79265-30-8
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR