Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.
Lieferant: Thermo Fisher Scientific
B21908.22
B21908.22EA
103
EUR
B21908.22
Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.
Ammonium iron(III) hexacyanoferrate(II) hydrate
CAS No.: 25869-00-5
Molecular Formula: (NH4)Fe[Fe(CN)6].xH2O
Formula Weight: 285.83(anhy)
MDL No.: MFCD01726733
Formel:
(NH₄)Fe[Fe(CN)₆]·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD01726733 CAS-Nummer: 25869-00-5 EINECS: 247-304-1 Merck Index: 14,7910 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...