(R)-(-)-1,2-Propandiol ≥98%
Lieferant: Thermo Fisher Scientific
Purity: 98%
CAS No.: 4254-14-2
Molecular Formula: C3H8O2
Formula Weight: 76.10
Flash Point: 107°(224°F)
Refractive Index: 1.4320
Boiling Point: 186-188°
Beilstein Reference: 1718872
Merck Reference: 14,7855
Optical Rotation: -16.5° (neat)
MDL No.: MFCD00066248
Formel:
C₃H₈O₂ Siedepunkt: 186…188 °C Dichte: 1,040 Flammpunkt: 107 °C (224 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066248 CAS-Nummer: 4254-14-2 Merck Index: 14,7855 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...