4'-Hydroxybiphenyl-4-carbonsäure ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4'-Hydroxy-4-biphenylcarboxylic acid, 4'-Hydroxy-[1,1'-biphenyl]-4-carboxylic acid
B21946.09
B21946.88
B21946.09EA
208
EUR
B21946.09
B21946.88
4'-Hydroxybiphenyl-4-carbonsäure ≥99%
4'-Hydroxybiphenyl-4-carbonsäure
Purity: 99%
CAS No.: 58574-03-1
Molecular Formula: C13H10O3
Formula Weight: 214.22
Melting Point: 292-295°
MDL No.: MFCD00059078
Formel:
C₁₃H₁₀O₃ Molecular Weight: 214.22 g/mol Schmelzpunkt: 292…295 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059078 CAS-Nummer: 58574-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...