4'-Hydroxybiphenyl-4-carbonsäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4'-Hydroxy-4-biphenylcarboxylic acid, 4'-Hydroxy-[1,1'-biphenyl]-4-carboxylic acid

B21946.09 B21946.88
B21946.09EA 208 EUR
B21946.09 B21946.88
4'-Hydroxybiphenyl-4-carbonsäure ≥99%
4'-Hydroxybiphenyl-4-carbonsäure

Purity: 99% CAS No.: 58574-03-1 Molecular Formula: C13H10O3 Formula Weight: 214.22 Melting Point: 292-295° MDL No.: MFCD00059078

Formel: C₁₃H₁₀O₃
Molecular Weight: 214.22 g/mol
Schmelzpunkt: 292…295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059078
CAS-Nummer: 58574-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR