2-Methyl-1-butanthiol ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Methylbutyl mercaptan, 1-Mercapto-2-methylbutane
B21949.06
B21949.14
B21949.06EA
51.2
EUR
B21949.06
B21949.14
2-Methyl-1-butanthiol ≥99%
2-Methyl-1-butanthiol
Purity: 99%
CAS No.: 1878-18-8
Molecular Formula: C5H12S
Formula Weight: 104.22
Flash Point: 19°(67°F)
Refractive Index: 1.4480
Boiling Point: 116-117°
UN No.: 1993
MDL No.: MFCD00004883
Formel:
C₅H₁₂S Siedepunkt: 116…117 °C Dichte: 0,845 Flammpunkt: 19 °C (67 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004883 CAS-Nummer: 1878-18-8 EINECS: 217-515-3 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...