4-Chlor-3,5-xylenol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B21964.22
B21964.36
B21964.22EA
22
EUR
B21964.22
B21964.36
4-Chlor-3,5-xylenol ≥98%
4-Chlor-3,5-xylenol
Purity: 98+%
CAS No.: 88-04-0
Molecular Formula: C8H9ClO
Formula Weight: 156.61
Flash Point: 138°(280°F)
Odour: Odourless
Boiling Point: ca 246°
Beilstein Reference: 1862539
Merck Reference: 14,2176
MDL No.: MFCD00002324
Formel:
C₈H₉ClO Siedepunkt: ∼246 °C Schmelzpunkt: 112…116 °C Dichte: 1,364 g/cm³ (20 °C) Flammpunkt: 138 °C (280 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002324 CAS-Nummer: 88-04-0 EINECS: 201-793-8 Merck Index: 14,2176 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...