4,4'-Diiodbiphenyl ≥99%

Lieferant: Thermo Fisher Scientific

B21970.14 B21970.22
B21970.14EA 46.9 EUR
B21970.14 B21970.22
4,4'-Diiodbiphenyl ≥99%
4,4'-Diiodbiphenyl

Purity: 99% CAS No.: 3001-15-8 Molecular Formula: C12H8I2 Formula Weight: 406.00 Melting Point: 205-211° UN No.: 3152 MDL No.: MFCD00001057

Formel: C₁₂H₈I₂
Schmelzpunkt: 205…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001057
CAS-Nummer: 3001-15-8
EINECS: 221-080-5
ADR: 9,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR