4,4'-Diiodbiphenyl ≥99%
Lieferant: Thermo Fisher Scientific
B21970.14
B21970.22
B21970.14EA
46.9
EUR
B21970.14
B21970.22
4,4'-Diiodbiphenyl ≥99%
4,4'-Diiodbiphenyl
Purity: 99%
CAS No.: 3001-15-8
Molecular Formula: C12H8I2
Formula Weight: 406.00
Melting Point: 205-211°
UN No.: 3152
MDL No.: MFCD00001057
Formel:
C₁₂H₈I₂ Schmelzpunkt: 205…211 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001057 CAS-Nummer: 3001-15-8 EINECS: 221-080-5 ADR: 9,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...