1-(4-Morpholino)cyclohexen ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 670-80-4
Molecular Formula: C10H17NO
Formula Weight: 167.25
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.5146
Boiling Point: 118-120°/10mm
Beilstein Reference: 118696
MDL No.: MFCD00006163
Formel:
C₁₀H₁₇NO Siedepunkt: 118…120 °C (10 mmHg) Dichte: 1,020 Flammpunkt: 68 °C (154 °F) |
MDL:
MFCD00006163 CAS-Nummer: 670-80-4 EINECS: 211-579-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...