trans-2,3-Epoxybutan ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

B22005.03 B22005.06 B22005.14
B22005.03EA 47.5 EUR
B22005.03 B22005.06 B22005.14
trans-2,3-Epoxybutan ≥97%
trans-2,3-Epoxybutane

Purity: 97% CAS No.: 21490-63-1 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -26°(-14°F) Refractive Index: 1.3730 Odour: Recognizable Boiling Point: 54-55° Beilstein Reference: 79772 UN No.: 3271 MDL No.: MFCD00005129

Formel: C₄H₈O
Siedepunkt: 54…55 °C
Dichte: 0,804
Flammpunkt: –26 °C (–14 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005129
CAS-Nummer: 21490-63-1
EINECS: 244-406-8
UN: 3271
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR