Hexakis(brommethyl)benzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1,2,3,4,5,6-Hexakis(bromomethyl)benzene
B22016.03
B22016.06
B22016.03EA
37.1
EUR
B22016.03
B22016.06
Hexakis(brommethyl)benzol ≥98%
Hexakis(brommethyl)benzol
Purity: 98%
CAS No.: 3095-73-6
Molecular Formula: C12H12Br6
Formula Weight: 635.65
Melting Point: ca 292° dec.
UN No.: 3261
MDL No.: MFCD00182539
Formel:
C₁₂H₁₂Br₆ Schmelzpunkt: ∼ 292 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00182539 CAS-Nummer: 3095-73-6 EINECS: 221-443-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...