2-Fluor-5-nitrobenzoesäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B22100.06
B22100.14
B22100.06EA
61.6
EUR
B22100.06
B22100.14
2-Fluor-5-nitrobenzoesäure ≥98%
2-Fluor-5-nitrobenzoesäure
Purity: 98%
CAS No.: 7304-32-7
Molecular Formula: C7H4FNO4
Formula Weight: 185.11
Melting Point: 142-144°
Beilstein Reference: 1912835
Formel:
C₇H₄FNO₄ Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134238 CAS-Nummer: 7304-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...