2-Fluor-5-nitrobenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B22100.06 B22100.14
B22100.06EA 61.6 EUR
B22100.06 B22100.14
2-Fluor-5-nitrobenzoesäure ≥98%
2-Fluor-5-nitrobenzoesäure

Purity: 98% CAS No.: 7304-32-7 Molecular Formula: C7H4FNO4 Formula Weight: 185.11 Melting Point: 142-144° Beilstein Reference: 1912835

Formel: C₇H₄FNO₄
Molecular Weight: 185.11 g/mol
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134238
CAS-Nummer: 7304-32-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR