2-Anilinoethanol 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 122-98-5
Molecular Formula: C6H5NH(CH2)2OH
Formula Weight: 137.18
Flash Point: 153°(307°F)
Refractive Index: 1.5790
Odour: Amine-like
pH: 10.2
Melting Point: -30°
Boiling Point: 284-286°
Beilstein Reference: 774672
UN No.: 2810
MDL No.: MFCD00002832
Formel:
C₆H₅NHCH₂CH₂OH Siedepunkt: 284…286 °C Schmelzpunkt: –30 °C Dichte: 1,087 Flammpunkt: 153 °C (307 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002832 CAS-Nummer: 122-98-5 EINECS: 204-588-1 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...