3-Fluorbrenzcatechin ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Fluorbenzol-1,2-diol
B22145.03
B22145.06
B22145.03EA
44.9
EUR
B22145.03
B22145.06
3-Fluorbrenzcatechin ≥99%
3-Fluorbrenzcatechin
Purity: 99%
CAS No.: 363-52-0
Molecular Formula: C6H5FO2
Formula Weight: 128.10
Melting Point: 71-73°
Beilstein Reference: 2436074
UN No.: 1759
MDL No.: MFCD00042582
Formel:
C₆H₅FO₂ Molecular Weight: 128.1 g/mol Siedepunkt: 218 Schmelzpunkt: 71…73 °C Dichte: 1.514 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042582 CAS-Nummer: 363-52-0 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...