Methyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}tyrosinat 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-Tyr-OMe, N-Boc-L-tyrosine Methyl Ester

B22164.06 B22164.14 B22164.22
B22164.06EA 63.3 EUR
B22164.06 B22164.14 B22164.22
Methyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}tyrosinat 99%
Boc-L-tyrosine methyl ester

Purity: 99% CAS No.: 4326-36-7 Molecular Formula: C15H21NO5 Formula Weight: 295.34 Melting Point: 105-108° Optical Rotation: +50° (c=1 chloroform) MDL No.: MFCD00191181

Formel: C₁₅H₂₁NO₅
Molecular Weight: 295.3 g/mol
Schmelzpunkt: 105…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191181
CAS-Nummer: 4326-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR