D(+)-Melezitose Hydrat ≥99%
Lieferant: Thermo Fisher Scientific
B22209.09
B22209.18
B22209.09EA
49.9
EUR
B22209.09
B22209.18
D(+)-Melezitose Hydrat ≥99%
D-(+)-Melezitose hydrate
Purity: 99%
CAS No.: 207511-10-2
Molecular Formula: C18H32O16.xH2O
Formula Weight: 504.46 (anhy)
Melting Point: ca 160° dec.
Beilstein Reference: 99539
Optical Rotation: +88°(c=4 in water)
MDL No.: MFCD00149448
Formel:
C₁₈H₃₄O₁₇ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149448 CAS-Nummer: 207511-10-2 EINECS: 209-894-9 Merck Index: 14,5819 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...