Sodium-1-propanesulphonate 99%

Lieferant: Thermo Fisher Scientific

B22210.03 B22210.06 B22210.14
B22210.03EA 29 EUR
B22210.03 B22210.06 B22210.14
Sodium-1-propanesulphonate 99%
Sodium-1-propanesulphonate

Purity: 99% CAS No.: 14533-63-2 Molecular Formula: CH3CH2CH2SO3Na Formula Weight: 146.14 MDL No.: MFCD00062546

Formel: C₃H₈O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00062546
CAS-Nummer: 14533-63-2
EINECS: 238-564-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR