Croconic acid disodium salt 97%

Lieferant: Thermo Fisher Scientific

B22235.01 B22235.03
B22235.01EA 45.6 EUR
B22235.01 B22235.03
Croconic acid disodium salt 97%
Croconic acid disodium salt

Purity: 97% CAS No.: 14379-00-1 Molecular Formula: C5Na2O5 Formula Weight: 186.03 Melting Point: 300° MDL No.: MFCD00191954

Formel: C₅Na₂O₅
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191954
CAS-Nummer: 14379-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR