1-(Difluormethoxy)-3-nitrobenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(Difluoromethoxy)nitrobenzene

B22267.14
B22267.14EA 156 EUR
B22267.14
1-(Difluormethoxy)-3-nitrobenzol ≥98%
1-(Difluormethoxy)-3-nitrobenzol

Purity: 98% CAS No.: 22236-07-3 Molecular Formula: C7H5F2NO3 Formula Weight: 189.12 Flash Point: 110°(230°F) Odour: Characteristic Boiling Point: 240-242° MDL No.: MFCD03407974

Formel: C₇H₅F₂NO₃
Molecular Weight: 189,12 g/mol
Siedepunkt: 240…242 °C
Dichte: 1,391
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03407974
CAS-Nummer: 22236-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR