Azo rubine

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Carmoisine B, acid Red 14, Brilliant Crimson Red, Sodium 4-hydroxy-3-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate

B22328.09 B22328.18 B22328.30
B22328.09EA 39.2 EUR
B22328.09 B22328.18 B22328.30
Azo rubine
Azo rubine

CAS No.: 3567-69-9 Molecular Formula: C20H12N2Na2O7S2 Formula Weight: 502.42 MDL No.: MFCD00003978

Formel: C₂₀H₁₄N₂O₇S₂
Molecular Weight: 502.42 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003978
CAS-Nummer: 3567-69-9
EINECS: 222-657-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR